This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01023070_m1
See other DAGLA GE Assays ›
Gene Symbol:
DAGLA
Gene Name:
diacylglycerol lipase alpha
Gene Aliases:
C11orf11, DAGL(ALPHA), DAGLALPHA, NSDDR
Chromosome Location:
Chr.11: 61680184 - 61747002 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01023070_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DAGLA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017018238.1 XP_016873727.1 1 - 2 319 124
XM_017018240.1 XP_016873729.1 1 - 2 319 124

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017018238.1, XM_017018240.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

neuroblast proliferation
arachidonic acid metabolic process
neurotransmitter biosynthetic process
diacylglycerol catabolic process
endocannabinoid signaling pathway
metal ion binding
acylglycerol lipase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0