This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01023302_m1
See other TTPAL GE Assays ›
Gene Symbol:
TTPAL
Gene Name:
alpha tocopherol transfer protein like
Gene Aliases:
C20orf121
Chromosome Location:
Chr.20: 44475886 - 44494603 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01023302_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TTPAL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039199.2 NP_001034288.1 3 - 4 761 87
NM_024331.4 NP_077307.2 4 - 5 785 87
XM_011529043.2 XP_011527345.1 4 - 5 897 87
XM_011529044.2 XP_011527346.1 4 - 5 1512 87
GenBank mRNA AK314045.1 - 3 - 4 720 87
BC003071.2 - 4 - 5 764 2989837 87
CR936621.1 - 3 - 4 715 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000007456.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000333.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TTPAL

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

transport
transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20004719.hg.1 ENST00000262605.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20004719.hg.1 ENST00000372904.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20004719.hg.1 NM_001039199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20004719.hg.1 NM_024331.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20004719.hg.1 TC20000333.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372906.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372906.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372906.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372906.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000372906.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000456317.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000456317.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000456317.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000456317.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000456317.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372906.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372906.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372906.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372906.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000372906.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000456317.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000456317.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000456317.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000456317.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000456317.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_001261839.1 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay TC2000007456.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 ENST00000456317.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 NM_001261839.1 Refseq
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 TC2000007456.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 ENST00000456317.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 ENST00000461134.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 NM_001261839.1 Refseq
Affymetrix Human Clariom D Assay PSR2000151743.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay PSR2000151741.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay PSR2000151741.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000151741.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay PSR2000151741.hg.1 NM_024331.4 Refseq
Affymetrix Human Clariom D Assay PSR2000151741.hg.1 TC2000007456.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000151741.hg.1 ENST00000262605.8 Ensembl
Affymetrix Human Clariom D Assay PSR2000151741.hg.1 ENST00000372904.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000151741.hg.1 NM_001039199.2 Refseq
Affymetrix Human Clariom D Assay PSR2000151741.hg.1 NM_024331.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0