This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01023897_m1
See other CDH1 GE Assays ›
Gene Symbol:
CDH1
Gene Name:
cadherin 1
Gene Aliases:
Arc-1, CD324, CDHE, ECAD, LCAM, UVO
Chromosome Location:
Chr.16: 68737290 - 68835542 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01023897_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317184.1 NP_001304113.1 8 - 9 1262 62
GenBank mRNA AB025105.1 - 8 - 9 1200 62

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
synapse assembly
response to toxic substance
single organismal cell-cell adhesion
pituitary gland development
negative regulation of cell-cell adhesion
extracellular matrix disassembly
extracellular matrix organization
neuron projection development
adherens junction organization
response to drug
positive regulation of transcription factor import into nucleus
positive regulation of transcription, DNA-templated
cellular response to lithium ion
cellular response to indole-3-methanol
protein localization to plasma membrane
establishment of protein localization to plasma membrane
glycoprotein binding
calcium ion binding
protein binding
beta-catenin binding
ankyrin binding
GTPase activating protein binding
gamma-catenin binding
cell adhesion molecule binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline