This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01025433_g1
See other LRIG2 GE Assays ›
Gene Symbol:
LRIG2
Gene Name:
leucine rich repeats and immunoglobulin like domains 2
Gene Aliases:
LIG-2, LIG2, UFS2
Chromosome Location:
Chr.1: 113073170 - 113132260 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01025433_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LRIG2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001312686.1 NP_001299615.1 6 - 7 1008 120
NM_014813.2 NP_055628.1 5 - 6 896 120
XM_005271369.2 XP_005271426.1 5 - 6 896 120
XM_017002952.1 XP_016858441.1 4 - 5 420 120
GenBank mRNA AB018349.1 - 5 - 6 857 120
AK291031.1 - 5 - 6 888 120
BC117368.1 - 5 - 6 720 40125919 120
BC117370.1 - 5 - 6 720 40125921 120
CR936669.1 - 6 - 7 999 120

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100009468.hg
This assay targets the same location as HTA transcript cluster(s) TC01000998.hg

Additional Information:

This assay is in the 5'UTR region of the XM_017002952.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LRIG2

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s)

sensory perception of sound
regulation of platelet-derived growth factor receptor signaling pathway
innervation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01015888.hg.1 TC01000998.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01015888.hg.1 ENST00000361127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01015888.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000470000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000470000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000470000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay PSR0100168194.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100168194.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay PSR0100168194.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100168194.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay PSR0100168194.hg.1 TC0100009468.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000361127.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466069.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000466161.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000470000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000470000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000470000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 ENST00000492207.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq
Affymetrix Human Clariom D Assay TC0100009468.hg.1 NM_014813.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0