This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01025523_m1
See other TTPAL GE Assays ›
Gene Symbol:
TTPAL
Gene Name:
alpha tocopherol transfer protein like
Gene Aliases:
C20orf121
Chromosome Location:
Chr.20: 44475886 - 44494603 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01025523_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TTPAL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039199.2 NP_001034288.1 1 - 2 105 76
NM_001261839.1 NP_001248768.1 1 - 2 105 76
XM_011529045.2 XP_011527347.1 1 - 2 105 76
GenBank mRNA AK314045.1 - 1 - 2 64 76
BP351602.1 - 1 - 2 107 76
BQ646722.1 - 1 - 2 71 76
CR936621.1 - 1 - 2 59 76

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK314045.1, CR936621.1, NM_001039199.2, NM_001261839.1, XM_011529045.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

transport
transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0