This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01025756_m1
See other EIF3A GE Assays ›
Gene Symbol:
EIF3A
Gene Name:
eukaryotic translation initiation factor 3 subunit A
Gene Aliases:
EIF3, EIF3S10, P167, TIF32, eIF3-p170, eIF3-theta, p180, p185
Chromosome Location:
Chr.10: 119035029 - 119080822 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01025756_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF3A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003750.2 NP_003741.1 11 - 12 1779 89
GenBank mRNA AK302575.1 - 11 - 12 1724 89
BC033769.1 - 11 - 12 1752 89
BC054342.1 - 11 - 12 1738 89
BC064926.1 - 11 - 12 1763 89
BC107776.1 - 11 - 12 1771 89
BC114429.1 - 11 - 12 1687 40027607 89
D50929.1 - 11 - 12 1761 89
JX312508.1 - 11 - 12 1780 89
U58046.1 - 11 - 12 1731 89
U78311.1 - 11 - 12 1746 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000011988.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001699.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EIF3A

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
formation of cytoplasmic translation initiation complex
translational initiation
regulation of translational initiation
IRES-dependent viral translational initiation
viral translational termination-reinitiation
translation initiation factor activity
structural molecule activity
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022372.hg.1 ENST00000369144.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022372.hg.1 ENST00000462527.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022372.hg.1 ENST00000541549.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022372.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022372.hg.1 TC10001699.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000182033.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000182033.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000182033.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000182033.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay PSR1000182033.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000182033.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000182033.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000182033.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay PSR1000182033.hg.1 TC1000011988.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0