This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01025757_m1
See other EIF3A GE Assays ›
Gene Symbol:
EIF3A
Gene Name:
eukaryotic translation initiation factor 3 subunit A
Gene Aliases:
EIF3, EIF3S10, P167, TIF32, eIF3-p170, eIF3-theta, p180, p185
Chromosome Location:
Chr.10: 119035029 - 119080822 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01025757_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF3A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003750.2 NP_003741.1 12 - 13 2121 87
GenBank mRNA AK302575.1 - 12 - 13 2066 87
AK307610.1 - 12 - 13 2099 87
BC054342.1 - 12 - 13 2080 87
BC114429.1 - 12 - 13 2029 40027607 87
D50929.1 - 12 - 13 2103 87
JX312508.1 - 12 - 13 2122 87
U58046.1 - 12 - 13 2073 87
U78311.1 - 12 - 13 2088 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000011988.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001699.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EIF3A

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
formation of cytoplasmic translation initiation complex
translational initiation
regulation of translational initiation
IRES-dependent viral translational initiation
viral translational termination-reinitiation
translation initiation factor activity
structural molecule activity
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022371.hg.1 ENST00000369144.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022371.hg.1 ENST00000462527.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022371.hg.1 ENST00000541549.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022371.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022371.hg.1 TC10001699.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10022369.hg.1 ENST00000369144.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022369.hg.1 ENST00000541549.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022369.hg.1 NM_003750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022369.hg.1 TC10001699.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000182032.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000182032.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000182032.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000182032.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay PSR1000182032.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000182032.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000182032.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000182032.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay PSR1000182032.hg.1 TC1000011988.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000462527.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay TC1000011988.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay PSR1000182030.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000182030.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000182030.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay PSR1000182030.hg.1 ENST00000369144.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000182030.hg.1 ENST00000541549.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000182030.hg.1 NM_003750.3 Refseq
Affymetrix Human Clariom D Assay PSR1000182030.hg.1 TC1000011988.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline