This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01025808_g1
See other CHD8 GE Assays ›
Gene Symbol:
CHD8
Gene Name:
chromodomain helicase DNA binding protein 8
Gene Aliases:
AUTS18, HELSNF1
Chromosome Location:
Chr.14: 21385194 - 21437298 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01025808_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHD8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001170629.1 NP_001164100.1 7 - 8 2090 150
NM_020920.3 NP_065971.2 8 - 9 1335 150
GenBank mRNA AB046784.3 - 7 - 8 1578 150
AK122910.1 - 8 - 9 1282 150
CR749315.1 - 8 - 9 1280 150

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400008631.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14000917.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CHD8

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
transcription, DNA-templated
brain development
covalent chromatin modification
negative regulation of Wnt signaling pathway
DNA duplex unwinding
ATP-dependent chromatin remodeling
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase III promoter
digestive tract development
canonical Wnt signaling pathway
negative regulation of canonical Wnt signaling pathway
negative regulation of fibroblast apoptotic process
p53 binding
DNA binding
DNA helicase activity
chromatin binding
protein binding
ATP binding
beta-catenin binding
DNA-dependent ATPase activity
methylated histone binding
histone binding
armadillo repeat domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR14010025.hg.1 ENST00000262707.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14010025.hg.1 ENST00000399982.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14010025.hg.1 ENST00000430710.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14010025.hg.1 ENST00000557364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14010025.hg.1 NM_001170629.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14010025.hg.1 NM_020920.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14010025.hg.1 TC14000917.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1400162378.hg.1 ENST00000399982.6 Ensembl
Affymetrix Human Clariom D Assay PSR1400162378.hg.1 ENST00000430710.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400162378.hg.1 ENST00000557364.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400162378.hg.1 NM_001170629.1 Refseq
Affymetrix Human Clariom D Assay PSR1400162378.hg.1 NM_020920.3 Refseq
Affymetrix Human Clariom D Assay PSR1400162378.hg.1 ENST00000399982.6 Ensembl
Affymetrix Human Clariom D Assay PSR1400162378.hg.1 ENST00000430710.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400162378.hg.1 ENST00000557364.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400162378.hg.1 NM_001170629.1 Refseq
Affymetrix Human Clariom D Assay PSR1400162378.hg.1 NM_020920.3 Refseq
Affymetrix Human Clariom D Assay PSR1400162378.hg.1 TC1400008631.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0