This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01026028_g1
See other CDKL4 GE Assays ›
Gene Symbol:
CDKL4
Gene Name:
cyclin dependent kinase like 4
Gene Aliases:
-
Chromosome Location:
Chr.2: 39174421 - 39246815 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01026028_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDKL4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011532815.2 XP_011531117.1 9 - 10 2349 92
GenBank mRNA AY847283.1 - 8 - 9 933 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
regulation of cell cycle
cyclin-dependent protein serine/threonine kinase activity
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000378803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000378803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000378803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000378803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000378803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000378803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000378803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000378803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000378803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000378803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000395035.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000395035.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000395035.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000395035.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000395035.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000395035.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000395035.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000395035.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000395035.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000419111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000419111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000419111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000419111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000451199.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000451199.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000451199.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 ENST00000451199.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 NM_001009565.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 NM_001009565.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 NM_001009565.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 NM_001009565.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 NM_001009565.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 NM_001009565.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 NM_001009565.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 NM_001009565.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 NM_001009565.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012318.hg.1 NM_001009565.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline