This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01026331_m1
See other SV2C GE Assays ›
Gene Symbol:
SV2C
Gene Name:
synaptic vesicle glycoprotein 2C
Gene Aliases:
-
Chromosome Location:
Chr.5: 75847463 - 76353939 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01026331_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SV2C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001297716.1 NP_001284645.1 3 - 4 962 80
NM_014979.3 NP_055794.3 3 - 4 962 80
XM_011543281.2 XP_011541583.1 3 - 4 1329 80
XM_011543282.2 XP_011541584.2 3 - 4 1889 80
GenBank mRNA AK094917.1 - 3 - 4 963 80
BC100824.1 - 3 - 4 898 40001296 80
BC100825.1 - 3 - 4 896 40001297 80
BC100826.1 - 3 - 4 896 40001302 80
BC100827.1 - 3 - 4 896 40001303 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500007820.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000366.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SV2C

Gene Ontology Categories:

Function(s) Process(es)

neurotransmitter transport
transmembrane transport
transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000506257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000506257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000506257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000581282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05005109.hg.1 TC05000366.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05005109.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05005109.hg.1 ENST00000502798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05005109.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay PSR0500153469.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500153469.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500153469.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay PSR0500153469.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay PSR0500153469.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500153469.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500153469.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay PSR0500153469.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay PSR0500153469.hg.1 TC0500007820.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000506257.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000506257.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000506257.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000581282.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0