This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01026336_m1
See other SV2C GE Assays ›
Gene Symbol:
SV2C
Gene Name:
synaptic vesicle glycoprotein 2C
Gene Aliases:
-
Chromosome Location:
Chr.5: 75847463 - 76353939 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01026336_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SV2C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001297716.1 NP_001284645.1 9 - 10 1705 162
NM_014979.3 NP_055794.3 9 - 10 1705 162
XM_011543281.2 XP_011541583.1 9 - 10 2072 162
XM_011543282.2 XP_011541584.2 9 - 10 2632 162
XM_017009244.1 XP_016864733.1 5 - 6 623 162
GenBank mRNA AB028977.1 - 7 - 8 766 162
AK094917.1 - 9 - 10 1706 162
BC100824.1 - 9 - 10 1641 40001296 162
BC100825.1 - 9 - 10 1639 40001297 162
BC100826.1 - 9 - 10 1639 40001302 162
BC100827.1 - 9 - 10 1639 40001303 162

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500007820.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000366.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SV2C

Gene Ontology Categories:

Function(s) Process(es)

neurotransmitter transport
transmembrane transport
transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05005119.hg.1 TC05000366.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05005119.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05005119.hg.1 ENST00000502798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05005119.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05005120.hg.1 TC05000366.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05005120.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05005120.hg.1 ENST00000502798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05005120.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000506257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000506257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000506257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000581282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000506257.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000506257.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000506257.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000581282.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay PSR0500153477.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500153477.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500153477.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay PSR0500153477.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay PSR0500153477.hg.1 ENST00000322285.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500153477.hg.1 ENST00000502798.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500153477.hg.1 NM_001297716.1 Refseq
Affymetrix Human Clariom D Assay PSR0500153477.hg.1 NM_014979.3 Refseq
Affymetrix Human Clariom D Assay PSR0500153477.hg.1 TC0500007820.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0