This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01026347_m1
See other RBM34 GE Assays ›
Gene Symbol:
RBM34
Gene Name:
RNA binding motif protein 34
Gene Aliases:
-
Chromosome Location:
Chr.1: 235131183 - 235161616 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01026347_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM34
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015014.2 NP_055829.2 4 - 5 628 101
NR_027762.1 3 - 4 491 101
XM_017000721.1 XP_016856210.1 4 - 5 988 101
GenBank mRNA AK292362.1 - 4 - 5 614 101
BC029451.1 - 4 - 5 628 4809263 101
CB052493.1 - 3 - 4 483 101

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RBM34

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Process(es)

nucleotide binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0