This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01026850_m1
See other GPRC6A GE Assays ›
Gene Symbol:
GPRC6A
Gene Name:
G protein-coupled receptor class C group 6 member A
Gene Aliases:
GPCR, bA86F4.3
Chromosome Location:
Chr.6: 116788512 - 116831128 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01026850_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPRC6A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286354.1 NP_001273283.1 1 - 2 234 143
NM_001286355.1 NP_001273284.1 1 - 2 234 143
NM_148963.3 NP_683766.2 1 - 2 234 143
XM_017010475.1 XP_016865964.1 2 - 3 2167 143
XM_017010476.1 XP_016865965.1 1 - 2 195 143
GenBank mRNA AF502962.1 - 1 - 2 212 143
AY435125.1 - 1 - 2 223 143
AY435126.1 - 1 - 2 223 143
AY435127.1 - 1 - 2 223 143
DY319308.1 - 1 - 2 250 143

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600012976.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06002053.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Ion Channel Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPRC6A

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
calcium-mediated signaling
response to amino acid
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0