This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01026851_m1
See other GPRC6A GE Assays ›
Gene Symbol:
GPRC6A
Gene Name:
G protein-coupled receptor class C group 6 member A
Gene Aliases:
GPCR, bA86F4.3
Chromosome Location:
Chr.6: 116788512 - 116831128 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01026851_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPRC6A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286354.1 NP_001273283.1 2 - 3 536 72
NM_001286355.1 NP_001273284.1 2 - 3 536 72
NM_148963.3 NP_683766.2 2 - 3 536 72
XM_017010475.1 XP_016865964.1 3 - 4 2469 72
XM_017010476.1 XP_016865965.1 2 - 3 497 72
GenBank mRNA AF502962.1 - 2 - 3 514 72
AW771483.1 - 1 - 2 72
AY435125.1 - 2 - 3 525 72
AY435126.1 - 2 - 3 525 72
AY435127.1 - 2 - 3 525 72
BG191110.1 - 1 - 2 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600012976.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06002053.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPRC6A

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
calcium-mediated signaling
response to amino acid
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline