This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01026853_m1
See other GPRC6A GE Assays ›
Gene Symbol:
GPRC6A
Gene Name:
G protein-coupled receptor class C group 6 member A
Gene Aliases:
GPCR, bA86F4.3
Chromosome Location:
Chr.6: 116788512 - 116831128 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01026853_m1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPRC6A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286354.1 NP_001273283.1 4 - 5 1065 127
NM_148963.3 NP_683766.2 4 - 5 1590 127
XM_017010475.1 XP_016865964.1 5 - 6 3523 127
XM_017010476.1 XP_016865965.1 4 - 5 1551 127
GenBank mRNA AF502962.1 - 4 - 5 1568 127
AY435125.1 - 4 - 5 1579 127
AY435126.1 - 4 - 5 1054 127

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600012976.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06002053.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Ion Channel 3' Most Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPRC6A

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
calcium-mediated signaling
response to amino acid
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000310357.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000368549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 ENST00000530250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_001286355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012976.hg.1 NM_148963.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cdktp:80/100.66.78.54:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline