This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01027424_m1
See other CHAF1A GE Assays ›
Gene Symbol:
CHAF1A
Gene Name:
chromatin assembly factor 1 subunit A
Gene Aliases:
CAF-1, CAF1, CAF1B, CAF1P150, P150
Chromosome Location:
Chr.19: 4402596 - 4448322 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01027424_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHAF1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005483.2 NP_005474.2 4 - 5 1120 75
XM_011527605.2 XP_011525907.1 4 - 5 1187 75
XM_011527606.2 XP_011525908.1 4 - 5 1187 75
GenBank mRNA BC041606.1 - 4 - 5 1045 75
BC052620.1 - 4 - 5 1119 75
BC067093.1 - 4 - 5 1121 6339027 75
U20979.1 - 4 - 5 1006 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900006691.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19000088.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CHAF1A

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA repair
DNA replication-dependent nucleosome assembly
protein complex assembly
cell cycle
chromatin assembly
chromatin binding
protein binding
identical protein binding
unfolded protein binding
chromo shadow domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000585854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000585854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000585854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000585854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000587368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000587368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000587368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000587580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000587580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000587580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000587580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000587580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000587739.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000587739.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000587739.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000587739.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000587739.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000589648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 ENST00000589648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19001411.hg.1 ENST00000301280.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19001411.hg.1 ENST00000344143.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19001411.hg.1 ENST00000535117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19001411.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19001411.hg.1 TC19000088.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19001410.hg.1 ENST00000301280.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19001410.hg.1 ENST00000344143.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19001410.hg.1 ENST00000535117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19001410.hg.1 NM_005483.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19001410.hg.1 TC19000088.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000585854.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000585854.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000585854.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000585854.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000587368.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000587368.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000587368.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000587580.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000587580.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000587580.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000587580.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000587580.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000587739.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000587739.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000587739.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000587739.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000587739.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000589648.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 ENST00000589648.1 Ensembl
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay TC1900006691.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay PSR1900146976.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay PSR1900146976.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900146976.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay PSR1900146976.hg.1 TC1900006691.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900146976.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay PSR1900146976.hg.1 ENST00000585371.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900146976.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay PSR1900146973.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay PSR1900146973.hg.1 ENST00000587739.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900146973.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay PSR1900146973.hg.1 TC1900006691.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900146973.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay PSR1900146973.hg.1 ENST00000587739.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900146973.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay PSR1900146977.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay PSR1900146977.hg.1 ENST00000587739.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900146977.hg.1 NM_005483.2 Refseq
Affymetrix Human Clariom D Assay PSR1900146977.hg.1 TC1900006691.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900146977.hg.1 ENST00000301280.9 Ensembl
Affymetrix Human Clariom D Assay PSR1900146977.hg.1 ENST00000587739.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900146977.hg.1 NM_005483.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlplw:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline