This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01027469_m1
See other RBM42 GE Assays ›
Gene Symbol:
RBM42
Gene Name:
RNA binding motif protein 42
Gene Aliases:
-
Chromosome Location:
Chr.19: 35629016 - 35637686 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01027469_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM42
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001319113.1 NP_001306042.1 2 - 3 414 67
NM_024321.4 NP_077297.2 2 - 3 414 67
XM_005259257.2 XP_005259314.1 2 - 3 409 67
GenBank mRNA AK301669.1 - 2 - 3 339 67
BC002868.2 - 2 - 3 353 3941786 67
BC004204.2 - 2 - 3 352 3532802 67
BC031682.1 - 2 - 3 381 5163682 67
HY031737.1 - 2 - 3 415 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900007869.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19000477.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RBM42

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of mRNA splicing, via spliceosome
nucleotide binding
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000262633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000586618.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000586618.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000586618.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000586618.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000586618.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000586618.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000586618.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000586618.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000588161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000588161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000588161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000588161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000588161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000588161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000588161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000588161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000588161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000588161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000588161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000588161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000588161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000588161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589559.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589559.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589559.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589559.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589559.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589559.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589559.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589559.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589559.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589559.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589559.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000589871.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592202.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 ENST00000592526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007869.hg.1 NM_024321.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rz6mw:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0