This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01029543_g1
See other PRSS56 GE Assays ›
Gene Symbol:
PRSS56
Gene Name:
protease, serine 56
Gene Aliases:
MCOP6
Chromosome Location:
Chr.2: 232520463 - 232525716 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01029543_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS56
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195129.1 NP_001182058.1 12 - 13 1660 56

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200011103.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001401.hg.1

Set Membership:

Amplicon length less than or equal to 70 Protease 3' Most Detect Genomic DNA Homo sapiens 1700 Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
camera-type eye development
serine-type endopeptidase activity

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3345309 - -
Whole Human Genome Oligo Microarray G4112A A_24_P15182 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02022120.hg.1 NM_001195129.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02022120.hg.1 TC02001401.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200179792.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179792.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179792.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179792.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179792.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179792.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179792.hg.1 TC0200011103.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200179793.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179793.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179793.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179793.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200179793.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200179793.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay PSR0200179793.hg.1 TC0200011103.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000449534.6 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000602410.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 ENST00000617714.1 Ensembl
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Affymetrix Human Clariom D Assay TC0200011103.hg.1 NM_001195129.1 Refseq
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A017103 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A017103 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 105205 - -
ABI Human Genome Survey Microarray Version 2.0 146592 - -
Array Name Probe IDs Targets Target Type
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1674447 NM_001195129.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1674447 CN280933.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1674447 NM_001195129.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1674447 CN280933.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-t8vjm:80/100.66.78.50:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline