This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01030668_m1
See other SLCO4C1 GE Assays ›
Gene Symbol:
SLCO4C1
Gene Name:
solute carrier organic anion transporter family member 4C1
Gene Aliases:
OATP-H, OATP-M1, OATP4C1, OATPX, PRO2176, SLC21A20
Chromosome Location:
Chr.5: 102233986 - 102296549 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01030668_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLCO4C1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_180991.4 NP_851322.3 7 - 8 1559 58
XM_011543370.1 XP_011541672.1 6 - 7 1237 58
XM_011543372.1 XP_011541674.1 9 - 10 1210 58
GenBank mRNA AF401643.1 - 7 - 8 1272 58
AK128854.1 - 7 - 8 1287 58
AY273896.2 - 7 - 8 1559 58
BX647349.1 - 7 - 8 1452 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500011596.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05001641.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLCO4C1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
spermatogenesis
cell differentiation
sodium-independent organic anion transport
sodium-independent organic anion transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05022295.hg.1 TC05001641.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05022295.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05022295.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay PSR0500180269.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500180269.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay PSR0500180269.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500180269.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay PSR0500180269.hg.1 TC0500011596.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 ENST00000310954.6 Ensembl
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq
Affymetrix Human Clariom D Assay TC0500011596.hg.1 NM_180991.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0