This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01030851_mH
See other SMARCA2 GE Assays ›
Gene Symbol:
SMARCA2
Gene Name:
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2
Gene Aliases:
BAF190, BRM, NCBRS, SNF2, SNF2L2, SNF2LA, SWI2, Sth1p, hBRM, hSNF2a
Chromosome Location:
Chr.9: 2015219 - 2193624 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01030851_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMARCA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289396.1 NP_001276325.1 27 - 28 4191 70
NM_001289397.1 NP_001276326.1 27 - 28 4030 70
NM_003070.4 NP_003061.3 27 - 28 4204 70
NM_139045.3 NP_620614.2 27 - 28 4204 70
GenBank mRNA AK296373.1 - 27 - 28 3592 70
AK299683.1 - 18 - 19 2397 70
X72889.1 - 27 - 28 4192 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000013.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SMARCA2

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
spermatid development
nervous system development
negative regulation of cell proliferation
negative regulation of cell growth
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription coactivator activity
chromatin binding
transcription coactivator activity
helicase activity
protein binding
ATP binding
DNA-dependent ATPase activity
histone binding
transcription regulatory region DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09000258.hg.1 TC09000013.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09000258.hg.1 ENST00000349721.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000258.hg.1 ENST00000382186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000258.hg.1 ENST00000382203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000258.hg.1 NM_003070.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09000256.hg.1 TC09000013.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09000256.hg.1 ENST00000302401.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000256.hg.1 ENST00000324954.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000256.hg.1 ENST00000349721.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000256.hg.1 ENST00000357248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000256.hg.1 ENST00000382183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000256.hg.1 ENST00000382185.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000256.hg.1 ENST00000382186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000256.hg.1 ENST00000382194.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000256.hg.1 ENST00000382203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000256.hg.1 NM_003070.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09000256.hg.1 NM_139045.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09000257.hg.1 TC09000013.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09000257.hg.1 ENST00000302401.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000257.hg.1 ENST00000324954.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000257.hg.1 ENST00000349721.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000257.hg.1 ENST00000357248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000257.hg.1 ENST00000382182.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000257.hg.1 ENST00000382183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000257.hg.1 ENST00000382185.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000257.hg.1 ENST00000382186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000257.hg.1 ENST00000382194.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000257.hg.1 ENST00000382203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000257.hg.1 NM_003070.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09000257.hg.1 NM_139045.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rz6mw:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0