Assay Details
| Gene Symbol: HDHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304510.1 | NP_001291439.1 | 1 - 2 | 287 | 83 | |
| GenBank mRNA | DA964284.1 | - | 1 - 2 | 264 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001304510.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000485934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000485934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000485934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | ENST00000485934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011259.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | ENST00000485934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | ENST00000485934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176740.hg.1 | TC0900011259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900176741.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176741.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176741.hg.1 | ENST00000485934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176741.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176741.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176741.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176741.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176741.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176741.hg.1 | ENST00000485934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176741.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176741.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176741.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176741.hg.1 | TC0900011259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | ENST00000485934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | ENST00000485934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176739.hg.1 | TC0900011259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000238379.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000374180.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000485934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000485934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000485934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | ENST00000485934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304509.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304510.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_001304511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_031219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011259.hg.1 | NM_031219.3 | Refseq |

HDHD3
Genomic Map