This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01032050_g1
See other P2RX1 GE Assays ›
Gene Symbol:
P2RX1
Gene Name:
purinergic receptor P2X 1
Gene Aliases:
P2X1
Chromosome Location:
Chr.17: 3896591 - 3916666 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01032050_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2RX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002558.3 NP_002549.1 10 - 11 1474 60
XM_006721529.2 XP_006721592.1 10 - 11 1263 60
XM_011523896.2 XP_011522198.1 11 - 12 1926 60
XM_011523897.2 XP_011522199.1 10 - 11 1374 60
XM_011523898.2 XP_011522200.1 10 - 11 1987 60
XM_011523899.2 XP_011522201.1 10 - 11 1978 60
GenBank mRNA AF020498.1 - 10 - 11 1309 60
AK313264.1 - 10 - 11 1273 60
BC027949.1 - 9 - 10 1556 60
BC044657.1 - 10 - 11 1249 5756349 60
U45448.1 - 10 - 11 1229 60
X83688.1 - 10 - 11 1206 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700009495.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17001014.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P2RX1

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

serotonin secretion by platelet
regulation of vascular smooth muscle contraction
transport
ion transport
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
signal transduction
insemination
blood coagulation
regulation of blood pressure
neuronal action potential
platelet activation
response to ATP
synaptic transmission, glutamatergic
purinergic nucleotide receptor signaling pathway
vasoconstriction
positive regulation of ion transport
ceramide biosynthetic process
protein homooligomerization
protein heterooligomerization
regulation of calcium ion transport
cation transmembrane transport
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
cation channel activity
ATP binding
drug binding
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000225538.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000571637.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000571637.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000571637.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000571637.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000572418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000576764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000576764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 ENST00000576764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009495.hg.1 NM_002558.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0