This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01032381_m1
See other ZNF239 GE Assays ›
Gene Symbol:
ZNF239
Gene Name:
zinc finger protein 239
Gene Aliases:
HOK-2, MOK2
Chromosome Location:
Chr.10: 43554516 - 43574619 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01032381_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF239
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001099284.1 NP_001092754.1 1 - 2 75 61
XM_011540234.2 XP_011538536.1 1 - 2 76 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000010446.hg
This assay targets the same location as HTA transcript cluster(s) TC10001203.hg

Additional Information:

This assay is in the 5'UTR region of the NM_001099284.1,XM_011540234.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
RNA binding
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10015115.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015115.hg.1 ENST00000535642.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015115.hg.1 NM_001099283.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015115.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015115.hg.1 TC10001203.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10015103.hg.1 ENST00000306006.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015103.hg.1 ENST00000339962.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015103.hg.1 ENST00000374446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015103.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015103.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015103.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015103.hg.1 NM_005674.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015103.hg.1 TC10001203.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10015113.hg.1 ENST00000339962.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015113.hg.1 ENST00000374446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015113.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015113.hg.1 ENST00000491188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015113.hg.1 ENST00000535642.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015113.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015113.hg.1 NM_001099283.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015113.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015113.hg.1 TC10001203.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000491188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000491188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015114.hg.1 ENST00000339962.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015114.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015114.hg.1 ENST00000491188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015114.hg.1 ENST00000535642.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015114.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015114.hg.1 NM_001099283.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015114.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015114.hg.1 TC10001203.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10015112.hg.1 ENST00000339962.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015112.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015112.hg.1 ENST00000535642.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10015112.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015112.hg.1 NM_001099283.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015112.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10015112.hg.1 TC10001203.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000171034.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Clariom D Assay PSR1000171034.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000171034.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000171034.hg.1 NM_001099282.1 Refseq
Affymetrix Human Clariom D Assay PSR1000171034.hg.1 NM_001099284.1 Refseq
Affymetrix Human Clariom D Assay PSR1000171034.hg.1 NM_005674.2 Refseq
Affymetrix Human Clariom D Assay PSR1000171034.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Clariom D Assay PSR1000171034.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000171034.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000171034.hg.1 NM_001099282.1 Refseq
Affymetrix Human Clariom D Assay PSR1000171034.hg.1 NM_001099284.1 Refseq
Affymetrix Human Clariom D Assay PSR1000171034.hg.1 NM_005674.2 Refseq
Affymetrix Human Clariom D Assay PSR1000171034.hg.1 TC1000010446.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000491188.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000491188.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_005674.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0