This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01032691_m1
See other ATXN1 GE Assays ›
Gene Symbol:
ATXN1
Gene Name:
ataxin 1
Gene Aliases:
ATX1, D6S504E, SCA1
Chromosome Location:
Chr.6: 16299112 - 16761490 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01032691_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATXN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC014548.1 - 2 - 3 268 121
BC039236.1 - 2 - 3 191 121
BC047894.1 - 2 - 3 297 121
BC063120.1 - 2 - 3 292 121
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CD521910.1 - 145 Chr. - - [-] on Build 121
GenBankmRNA BU602093.1 - 2 - 3 291 Chr. 6 - 16739475 - 16761469 [-] on Build GRCh38 121
GenBankmRNA HY026825.1 - 2 - 3 285 Chr. 6 - 16739717 - 16761463 [-] on Build GRCh38 121
GenBankmRNA DB202047.1 - 2 - 3 281 Chr. 6 - 16739611 - 16761460 [-] on Build GRCh38 121
GenBankmRNA BG253572.1 - 2 - 3 295 Chr. 6 - 16739511 - 16761475 [-] on Build GRCh38 121
GenBankmRNA BM919920.1 - 2 - 3 293 Chr. 6 - 16739237 - 16761439 [-] on Build GRCh38 121
GenBankmRNA DB103131.1 - 2 - 3 281 Chr. 6 - 16739637 - 16761460 [-] on Build GRCh38 121
GenBankmRNA DA415596.1 - 3 - 4 332 Chr. 6 - 16739602 - 16761459 [-] on Build GRCh38 121
GenBankmRNA BG497560.1 - 2 - 3 189 Chr. 6 - 16739282 - 16761371 [-] on Build GRCh38 121
GenBankmRNA DA072514.1 - 3 - 4 333 Chr. 6 - 16739686 - 16761463 [-] on Build GRCh38 121
GenBankmRNA DA975674.1 - 1 - 2 130 Chr. 6 - 16739485 - 16753347 [-] on Build GRCh38 122
GenBankmRNA DA884819.1 - 2 - 3 281 Chr. 6 - 16739648 - 16761460 [-] on Build GRCh38 121
GenBankmRNA DB035376.1 - 2 - 3 284 Chr. 6 - 16739644 - 16761463 [-] on Build GRCh38 121
GenBankmRNA DA544691.1 - 2 - 3 280 Chr. 6 - 16739653 - 16761459 [-] on Build GRCh38 121
GenBankmRNA DA014569.1 - 2 - 3 290 Chr. 6 - 16739626 - 16761469 [-] on Build GRCh38 121

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream guanosine monophosphate reductase GMPR GMPR 1,
GMPR1
-444.0kb Anti-sense Chr. 6 - 16238580 - 16295549 on NCBI Build GRCh38
Downstream uncharacterized LOC101928433 LOC101928433 6.7kb Anti-sense Chr. 6 - 16761138 - 16766883 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000467008.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000467008.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000467008.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000467008.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000467008.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000467008.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000479680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000479680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000479680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000479680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000483591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000483591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000483591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000483591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000483591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000483591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000483591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000483954.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000483954.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000492857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000492857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000492857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000492857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000492857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000495178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000495178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000495178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000498374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000498374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000498374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 ENST00000626941.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000467008.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000467008.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000467008.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000467008.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000467008.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000467008.5 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000479680.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000479680.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000479680.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000479680.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000483591.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000483591.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000483591.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000483591.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000483591.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000483591.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000483591.6 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000483954.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000483954.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000492857.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000492857.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000492857.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000492857.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000492857.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000495178.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000495178.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000495178.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000498374.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000498374.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000498374.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 ENST00000626941.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay TC0600010921.hg.1 NM_001128164.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline