This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01032884_m1
See other RAC2 GE Assays ›
Gene Symbol:
RAC2
Gene Name:
ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2)
Gene Aliases:
EN-7, Gx, HSPC022, p21-Rac2
Chromosome Location:
Chr.22: 37225261 - 37244299 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01032884_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002872.4 NP_002863.1 6 - 7 726 80
GenBank mRNA BC001485.2 - 6 - 7 698 3509104 80
BC018735.1 - 2 - 3 245 80
BI908751.1 - 6 - 7 729 80
CR456555.1 - 6 - 7 608 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200008641.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000725.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Human Phagocytosis of Microbes Human NGF Pathway HUMAN INSULIN RECEPTOR PATHWAY HUMAN BREAST CANCER REGULATION BY STATHMIN1 HUMAN GPCR PATHWAY HUMAN GUIDANCE CUES AND GROWTH CONE MOTILITY HUMAN RHO FAMILY GTPASES HUMAN RAS PATHWAY HUMAN PANCREATIC ADENOCARCINOMA Inventoried Human_Phagocytosis_of_Microbes Human_NGF_Pathway Human_Pancreatic_Adenocarcinoma Human_Guidance_Cues_and_Growth_Cone_Motility Human_GPCR_Pathway Human_Insulin_Receptor_Pathway Human_Ras_Pathway Human_Rho_Family_GTPases Human_Breast_Cancer_Regulation_by_Stathmin1 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RAC2

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
actin filament organization
signal transduction
G-protein coupled receptor signaling pathway
small GTPase mediated signal transduction
positive regulation of cell proliferation
regulation of hydrogen peroxide metabolic process
positive regulation of lamellipodium assembly
regulation of cell-substrate adhesion
cell projection assembly
platelet activation
regulation of T cell proliferation
regulation of mast cell degranulation
bone resorption
regulation of protein kinase activity
regulation of small GTPase mediated signal transduction
regulation of respiratory burst
regulation of mast cell chemotaxis
lymphocyte aggregation
positive regulation of neutrophil chemotaxis
regulation of neutrophil migration
positive regulation of protein targeting to mitochondrion
GTPase activity
GTP binding
protein kinase regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000401529.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000401529.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000401529.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000401529.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000441619.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000441619.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000441619.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000441619.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000441619.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000441619.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000441619.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000469532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000469532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000469532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000469532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000469532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000481215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000481215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000481215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000481215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000481215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000481215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0