| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000383681.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000383681.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000383681.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000383681.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000383681.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000383681.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000478595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000478595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000478595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000478595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000478595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000478595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000498096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000498096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000498096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000498096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000498096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000498096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000607516.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000607516.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000607516.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000607516.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
ENST00000607516.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03010385.hg.1 |
ENST00000473539.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03010385.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03010385.hg.1 |
TC03000572.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300205051.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300205051.hg.1 |
ENST00000607516.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300205051.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300205051.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300205051.hg.1 |
ENST00000607516.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300205051.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300205051.hg.1 |
TC0300013859.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000383681.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000383681.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000383681.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000383681.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000383681.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000383681.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000478595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000478595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000478595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000478595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000478595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000478595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000498096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000498096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000498096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000498096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000498096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000498096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000607516.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000607516.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000607516.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000607516.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
ENST00000607516.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013859.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
ENST00000383681.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
ENST00000478595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
ENST00000498096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
ENST00000315711.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
ENST00000383681.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
ENST00000473539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
ENST00000478595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
ENST00000498096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
NM_001004196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
NM_005944.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300205054.hg.1 |
TC0300013859.hg.1 |
Clariom D TC |