This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01033710_m1
See other PNPLA4 GE Assays ›
Gene Symbol:
PNPLA4
Gene Name:
patatin like phospholipase domain containing 4
Gene Aliases:
DXS1283E, GS2, iPLA2eta
Chromosome Location:
Chr.X: 7898763 - 7928587 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01033710_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PNPLA4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011545588.1 XP_011543890.1 1 - 2 915 108
GenBank mRNA BM850121.1 - 1 - 2 67 108
DC421932.1 - 1 - 2 67 108

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011545588.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

hydrolase lipase phospholipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
triglyceride catabolic process
retinol metabolic process
cellular lipid metabolic process
lipid homeostasis
molecular_function
triglyceride lipase activity
retinyl-palmitate esterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000442940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000442940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000442940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000442940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000442940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000442940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000381042.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000442940.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000442940.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000442940.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000442940.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000442940.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000442940.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000444736.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 ENST00000537427.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001142389.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_001172672.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_004650.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009018.hg.1 NM_004650.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0