This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01035589_m1
See other IGSF3 GE Assays ›
Gene Symbol:
IGSF3
Gene Name:
immunoglobulin superfamily member 3
Gene Aliases:
EWI-3, LCDD, V8
Chromosome Location:
Chr.1: 116574398 - 116667755 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01035589_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007237.2 NP_001007238.1 6 - 7 2389 63
NM_001542.3 NP_001533.2 6 - 7 2311 63
XM_005270793.2 XP_005270850.1 7 - 8 2421 63
XM_005270794.4 XP_005270851.1 5 - 6 1659 63
XM_006710593.3 XP_006710656.1 6 - 7 1722 63
XM_011541315.1 XP_011539617.1 7 - 8 2473 63
GenBank mRNA AB007935.2 - 5 - 6 1721 63
AF031174.1 - 6 - 7 1786 63
AK097809.1 - 2 - 3 721 63
BC110651.1 - 6 - 7 2389 6166346 63
BC130279.1 - 5 - 6 2059 40146310 63
BC144132.1 - 5 - 6 2059 9052652 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100015401.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003030.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGSF3

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

lacrimal gland development
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000481589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000481589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008186.hg.1 ENST00000625078.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0