This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01035590_m1
See other IGSF3 GE Assays ›
Gene Symbol:
IGSF3
Gene Name:
immunoglobulin superfamily member 3
Gene Aliases:
EWI-3, LCDD, V8
Chromosome Location:
Chr.1: 116574398 - 116667755 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01035590_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007237.2 NP_001007238.1 7 - 8 2797 150
NM_001542.3 NP_001533.2 7 - 8 2719 150
XM_005270793.2 XP_005270850.1 8 - 9 2829 150
XM_005270794.4 XP_005270851.1 6 - 7 2067 150
XM_006710593.3 XP_006710656.1 7 - 8 2130 150
XM_011541315.1 XP_011539617.1 8 - 9 2881 150
XM_011541316.2 XP_011539618.1 2 - 3 1610 150
GenBank mRNA AB007935.2 - 6 - 7 2129 150
AF031174.1 - 7 - 8 2194 150
AK097809.1 - 3 - 4 1129 150
BC110651.1 - 7 - 8 2797 6166346 150
BC130279.1 - 6 - 7 2467 40146310 150
BC144132.1 - 6 - 7 2467 9052652 150

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100015401.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003030.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGSF3

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

lacrimal gland development
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000481589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000481589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008186.hg.1 ENST00000625078.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008186.hg.1 ENST00000625078.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0