This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01035593_m1
See other IGSF3 GE Assays ›
Gene Symbol:
IGSF3
Gene Name:
immunoglobulin superfamily member 3
Gene Aliases:
EWI-3, LCDD, V8
Chromosome Location:
Chr.1: 116574398 - 116667755 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01035593_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001542.3 NP_001533.2 4 - 5 1857 139
XM_005270793.2 XP_005270850.1 5 - 6 1967 139
XM_006710593.3 XP_006710656.1 4 - 5 1268 139
XM_011541315.1 XP_011539617.1 5 - 6 2019 139
GenBank mRNA AF031174.1 - 4 - 5 1332 139

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100015401.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGSF3

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

lacrimal gland development
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000318837.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000369486.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000481589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 ENST00000481589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001007237.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015401.hg.1 NM_001542.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0