This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01035653_m1
See other LIMA1 GE Assays ›
Gene Symbol:
LIMA1
Gene Name:
LIM domain and actin binding 1
Gene Aliases:
EPLIN, SREBP3
Chromosome Location:
Chr.12: 50175780 - 50283570 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01035653_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIMA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113546.1 NP_001107018.1 8 - 9 1211 59
NM_001113547.1 NP_001107019.1 5 - 6 1092 59
XM_011538455.2 XP_011536757.1 8 - 9 1271 59
XM_017019429.1 XP_016874918.1 3 - 4 721 59
GenBank mRNA AB209512.1 - 8 - 9 1147 59
AF198455.1 - 5 - 6 1030 59

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of actin filament depolymerization
ruffle organization
actin filament bundle assembly
cell-cell adhesion
actin monomer binding
protein binding
zinc ion binding
actin filament binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0