This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01035816_m1
See other ENPP5 GE Assays ›
Gene Symbol:
ENPP5
Gene Name:
ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative)
Gene Aliases:
NPP-5, NPP5
Chromosome Location:
Chr.6: 46159182 - 46171045 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01035816_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ENPP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290072.1 NP_001277001.1 4 - 5 1308 84
NM_001290073.1 NP_001277002.1 3 - 4 955 84
NM_021572.5 NP_067547.1 3 - 4 1246 84
XM_005249260.3 XP_005249317.1 3 - 4 1145 84
XM_011514786.2 XP_011513088.1 4 - 5 1163 84
GenBank mRNA AK295192.1 - 3 - 4 916 84
AY358514.1 - 4 - 5 1251 84
BC027615.2 - 3 - 4 1146 4839342 84
BX647968.1 - 3 - 4 1217 84

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ENPP5

Gene Ontology Categories:

Function(s) Process(es)

cell communication
metabolic process
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline