This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01036309_m1
See other NDUFV3 GE Assays ›
Gene Symbol:
NDUFV3
Gene Name:
NADH:ubiquinone oxidoreductase subunit V3
Gene Aliases:
CI-10k, CI-9KD
Chromosome Location:
Chr.21: 42893268 - 42911396 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01036309_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFV3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021075.3 NP_066553.3 3 - 4 1335 106
GenBank mRNA AK056041.1 - 5 - 6 1792 106
AK308228.1 - 3 - 4 1292 106
BC021217.2 - 3 - 4 1322 4302265 106
BU629860.1 - 1 - 2 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100008529.hg.1
This assay targets the same location as HTA transcript cluster(s) TC21001063.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDUFV3

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, NADH to ubiquinone
mitochondrial respiratory chain complex I assembly
NADH dehydrogenase (ubiquinone) activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000340344.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000340344.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000340344.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000340344.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000340344.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000340344.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000340344.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000354250.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000354250.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000354250.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000354250.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000354250.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000354250.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000354250.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000460740.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 ENST00000460740.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 NM_001001503.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 NM_001001503.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 NM_001001503.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 NM_001001503.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 NM_001001503.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 NM_001001503.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 NM_021075.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 NM_021075.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 NM_021075.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 NM_021075.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 NM_021075.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 NM_021075.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 NM_021075.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008529.hg.1 NM_021075.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-l2zpb:80/100.66.75.138:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0