This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01036787_m1
See other NFIB GE Assays ›
Gene Symbol:
NFIB
Gene Name:
nuclear factor I B
Gene Aliases:
CTF, HMGIC/NFIB, NF-I/B, NF1-B, NFI-B, NFI-RED, NFIB2, NFIB3
Chromosome Location:
Chr.9: 14081843 - 14398983 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01036787_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NFIB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282787.1 NP_001269716.1 9 - 10 1050 65
XM_005251467.2 XP_005251524.1 10 - 11 2123 65
XM_005251468.4 XP_005251525.1 10 - 11 1596 65
XM_005251469.2 XP_005251526.1 10 - 11 2096 65
XM_005251471.3 XP_005251528.1 9 - 10 2958 65
XM_006716773.2 XP_006716836.1 10 - 11 2120 65
GenBank mRNA BX648416.1 - 9 - 10 1047 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900009569.hg.1

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

chondrocyte differentiation
DNA replication
transcription from RNA polymerase II promoter
glial cell differentiation
principal sensory nucleus of trigeminal nerve development
anterior commissure morphogenesis
hindbrain development
positive regulation of transcription from RNA polymerase II promoter
Clara cell differentiation
Type I pneumocyte differentiation
Type II pneumocyte differentiation
salivary gland cavitation
cell differentiation involved in salivary gland development
lung ciliated cell differentiation
commissural neuron axon guidance
negative regulation of pri-miRNA transcription from RNA polymerase II promoter
negative regulation of mesenchymal cell proliferation involved in lung development
negative regulation of epithelial cell proliferation involved in lung morphogenesis
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0900165548.hg.1 ENST00000397575.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900165548.hg.1 ENST00000397579.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900165548.hg.1 ENST00000397581.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900165548.hg.1 ENST00000543693.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900165548.hg.1 NM_001282787.1 Refseq
Affymetrix Human Clariom D Assay PSR0900165548.hg.1 ENST00000397575.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900165548.hg.1 ENST00000397579.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900165548.hg.1 ENST00000397581.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900165548.hg.1 ENST00000543693.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900165548.hg.1 NM_001282787.1 Refseq
Affymetrix Human Clariom D Assay PSR0900165548.hg.1 TC0900009569.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline