This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01037089_g1
See other UNG GE Assays ›
Gene Symbol:
UNG
Gene Name:
uracil DNA glycosylase
Gene Aliases:
DGU, HIGM4, HIGM5, UDG, UNG1, UNG15, UNG2
Chromosome Location:
Chr.12: 109097594 - 109110993 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01037089_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003362.3 NP_003353.1 1 - 2 516 133
NM_080911.2 NP_550433.1 2 - 3 420 133
GenBank mRNA AK291341.1 - 1 - 2 440 133
AK299416.1 - 2 - 3 360 133
AK313552.1 - 2 - 3 417 133
BC015205.1 - 1 - 2 399 3913667 133
BC050634.2 - 1 - 2 410 6500377 133
BG326541.1 - 1 - 2 450 132
BM928006.1 - 471 133
BQ950839.1 - 2 - 3 431 133
CR541673.1 - 1 - 2 307 133
CR749342.1 - 1 - 2 484 133
X15653.1 - 1 - 2 413 133
Y09008.1 - 2 - 3 404 133

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200008792.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000843.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UNG

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
base-excision repair
viral process
somatic hypermutation of immunoglobulin genes
somatic recombination of immunoglobulin gene segments
negative regulation of apoptotic process
depyrimidination
positive regulation of isotype switching
base-excision repair, AP site formation via deaminated base removal
damaged DNA binding
uracil DNA N-glycosylase activity
protein binding
enzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000540158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000540158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000540158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000540158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009838.hg.1 ENST00000603734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12010802.hg.1 ENST00000336865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12010802.hg.1 ENST00000537518.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12010802.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12010802.hg.1 TC12000843.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12010805.hg.1 ENST00000336865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12010805.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12010805.hg.1 TC12000843.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12010808.hg.1 ENST00000242576.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12010808.hg.1 ENST00000336865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12010808.hg.1 ENST00000537518.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12010808.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12010808.hg.1 NM_080911.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12010808.hg.1 TC12000843.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12010804.hg.1 ENST00000336865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12010804.hg.1 ENST00000537518.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12010804.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12010804.hg.1 TC12000843.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12010801.hg.1 ENST00000336865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12010801.hg.1 NM_003362.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12010801.hg.1 TC12000843.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1400009838.hg.1 ENST00000603734.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200164954.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200164954.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200164954.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200164954.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay PSR1200164954.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200164954.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200164954.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200164954.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay PSR1200164954.hg.1 TC1200008792.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000540158.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000540158.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000540158.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000540158.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
Affymetrix Human Clariom D Assay PSR1200164952.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200164952.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay PSR1200164952.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200164952.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay PSR1200164952.hg.1 TC1200008792.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 ENST00000540158.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 NM_080911.2 Refseq
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 ENST00000242576.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 ENST00000540158.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 NM_080911.2 Refseq
Affymetrix Human Clariom D Assay PSR1200164959.hg.1 TC1200008792.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200164956.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200164956.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200164956.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200164956.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay PSR1200164956.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200164956.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200164956.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200164956.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay PSR1200164956.hg.1 TC1200008792.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200164957.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200164957.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200164957.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200164957.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay PSR1200164957.hg.1 ENST00000336865.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200164957.hg.1 ENST00000446767.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200164957.hg.1 ENST00000539287.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200164957.hg.1 NM_003362.3 Refseq
Affymetrix Human Clariom D Assay PSR1200164957.hg.1 TC1200008792.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline