| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000540158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000540158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000540158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
ENST00000540158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12010810.hg.1 |
ENST00000242576.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12010810.hg.1 |
ENST00000336865.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12010810.hg.1 |
ENST00000537518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12010810.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12010810.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12010810.hg.1 |
TC12000843.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000540158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000540158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000540158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
ENST00000540158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008792.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200164964.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200164964.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200164964.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200164964.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200164964.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200164964.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200164964.hg.1 |
ENST00000242576.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200164964.hg.1 |
ENST00000336865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200164964.hg.1 |
ENST00000446767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200164964.hg.1 |
ENST00000539287.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200164964.hg.1 |
NM_003362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200164964.hg.1 |
NM_080911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200164964.hg.1 |
TC1200008792.hg.1 |
Clariom D TC |