This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01037527_s1
See other HDHD3 GE Assays ›
Gene Symbol:
HDHD3
Gene Name:
haloacid dehalogenase like hydrolase domain containing 3
Gene Aliases:
2810435D12Rik, C9orf158
Chromosome Location:
Chr.9: 113373416 - 113377009 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01037527_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDHD3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001304509.1 NP_001291438.1 3 1443 77
NM_001304510.1 NP_001291439.1 3 1402 77
NM_001304511.1 NP_001291440.1 2 1328 77
NM_031219.3 NP_112496.1 2 1656 77
GenBank mRNA AJ420424.1 - 2 1159 77
BC005048.2 - 1 1047 3615096 77
BC031878.1 - 2 1656 4747097 77

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HDHD3

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
protein binding
hydrolase activity

MicroArray Probes:

Agilent Affymetrix ArrayStar Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P27251059 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06907006 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06906995 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27251052 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06906996 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06907014 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27251044 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06907013 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27251057 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27251061 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27251070 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06906997 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27251062 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06907016 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06907018 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06907003 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27251080 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27251063 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27251076 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27251072 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06907012 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27251056 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06907011 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27251069 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06907002 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06907021 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06906999 - -
Agilent SurePrint G3 Human CGH Microarray 4x180K 12274 - -
Agilent SurePrint G3 Human CGH Microarray 8x60K 6153 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 250360 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_23_P112548 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P112548 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 2340 - -
Agilent Human Genome CGH Microarray 44B A_14_P138008 - -
Agilent Human Genome CGH Microarray 44A A_14_P138008 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 29129 - -
Agilent Human Genome CGH + SNP Microarray A_18_P16976845 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_23_P112548 - -
SurePrint G3 Human GE Microarray G4851A A_23_P112548 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907002 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907009 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251044 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907012 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251063 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907006 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251069 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06906994 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907013 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06906999 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251059 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251061 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907016 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251070 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251072 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06906995 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251057 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907018 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06906997 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907011 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907019 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907014 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06906996 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907003 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907022 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251076 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251080 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251052 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251056 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06906996 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251063 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907003 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06906994 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251070 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907002 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907012 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251080 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907018 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06906999 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06906995 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907011 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907014 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251056 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907019 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251057 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907013 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06906997 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907022 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251072 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251076 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251061 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907009 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251052 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251044 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251069 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907016 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27251059 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06907006 - -
Whole Human Genome Oligo Microarray G4112A A_23_P112548 - -
Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P138008 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06907019 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06907013 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06907010 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06906995 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06907002 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06907016 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06907005 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06906994 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06907012 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06906999 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06907017 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06906997 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06907014 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06907006 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06907020 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06907022 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06907008 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P06907013 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16255295 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P06907014 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16255296 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P138008 - -
Agilent Human Genome CGH Microarray 4x44K A_14_P138008 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P112548 - -
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 72461 - -
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 60074 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P138008 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06907020 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06907002 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06907016 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06907005 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06907010 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16255298 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16255295 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06907008 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06907006 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06907022 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06906995 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06906997 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06907012 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06906999 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06907019 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06906994 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06907014 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P138008 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array NM_031219_at - -
Affymetrix Human Gene 1.0 ST Array 8163506 - -
Affymetrix Human Gene 1.0 ST Array 8163507 - -
Affymetrix Human Gene 1.0 ST Array 8163508 - -
Affymetrix Human Gene 1.1 ST Array 8163505 - -
Affymetrix HT HG-U133+ PM Array Plate 221256_PM_s_at - -
Affymetrix Human Genome U219 Array 11754310_PM_a_at - -
Affymetrix Human Genome U219 Array 11754310_a_at - -
Human Genome U133 Plus 2.0 Array 221256_s_at - -
Human Genome U133 Plus 2.0 Array 221256_PM_s_at - -
Human Genome U133A 2.0 Array 221256_s_at - -
Human Genome U133A 2.0 Array 221256_PM_s_at - -
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_031219_at - -
AffymetrixPathwork Diagnostics Pathchip pw582207 - -
Affymetrix Human Genome U133 Plus 2.0 Array 221256_s_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 221256_s_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 221256_s_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 221256_s_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 221256_s_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 221256_s_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 221256_s_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 221256_s_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 221256_s_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 221256_s_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 221256_s_at2 - -
Human Genome U95 Set 46277_at - -
Human Genome U95 Set 46277_PM_at - -
Human Genome U133 Set 221256_PM_s_at - -
Human Genome U133 Set 221256_s_at - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A037933 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A037933 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_1726466 CR609038.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1726466 CR620220.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1726466 CR590490.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1726466 CR602117.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1726466 CR622772.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1726466 CR609038.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1726466 CR620220.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1726466 CR590490.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1726466 CR602117.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1726466 CR622772.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1726466 CR609038.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1726466 CR620220.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1726466 CR590490.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1726466 CR602117.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1726466 CR622772.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1726466 CR609038.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1726466 CR620220.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1726466 CR590490.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1726466 CR602117.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1726466 CR622772.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1726466 CR609038.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1726466 CR620220.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1726466 CR590490.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1726466 CR602117.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1726466 CR622772.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_31377665-S CR609038.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_31377665-S CR620220.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_31377665-S CR590490.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_31377665-S CR602117.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_31377665-S CR622772.1 human_transcriptome
Illumina HumanRef-8 v2 Expression BeadChip GI_14769-S - -
Illumina human-6 v1.0 expression beadchip 0006110064 - -
Illumina humanRef-8 v1.0 expression beadchip 0006110064 - -
Illumina human-6 v2.0 expression beadchip ILMN_1726466 CR609038.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1726466 CR620220.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1726466 CR590490.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1726466 CR602117.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1726466 CR622772.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1726466 CR609038.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1726466 CR620220.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1726466 CR590490.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1726466 CR602117.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1726466 CR622772.1 human_transcriptome
Illumina Sentrix Human-6 v2 Expression BeadChip 7380377 - -
Illumina HumanRef-6 v2.0 expression beadchip ILMN_14769 - -
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1726466 CR609038.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1726466 CR620220.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1726466 CR590490.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1726466 CR602117.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1726466 CR622772.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1726466 CR609038.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1726466 CR620220.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1726466 CR590490.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1726466 CR602117.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1726466 CR622772.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1726466 CR609038.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1726466 CR620220.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1726466 CR590490.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1726466 CR602117.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1726466 CR622772.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1726466 CR609038.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1726466 CR620220.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1726466 CR590490.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1726466 CR602117.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1726466 CR622772.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1726466 CR609038.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1726466 CR620220.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1726466 CR590490.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1726466 CR602117.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1726466 CR622772.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline