This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01037715_m1
See other CHST15 GE Assays ›
Gene Symbol:
CHST15
Gene Name:
carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15
Gene Aliases:
BRAG, GALNAC4S-6ST
Chromosome Location:
Chr.10: 124006060 - 124093684 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01037715_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 330.60
330.6
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHST15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270764.1 NP_001257693.1 1 - 2 133 103
NM_001270765.1 NP_001257694.1 1 - 2 133 103
XM_005269892.4 XP_005269949.1 1 - 2 210 103
XM_005269893.3 XP_005269950.1 1 - 2 210 103
XM_005269894.4 XP_005269951.1 1 - 2 210 103
XM_011539857.1 XP_011538159.1 1 - 2 210 103
GenBank mRNA AK299546.1 - 1 - 2 124 103
BC050540.1 - 1 - 2 113 5741278 103
CR749804.1 - 1 - 2 216 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK299546.1, BC050540.1, CR749804.1, NM_001270764.1, NM_001270765.1, XM_005269892.4, XM_005269893.3, XM_005269894.4, XM_011539857.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

hexose biosynthetic process
chondroitin sulfate biosynthetic process
3'-phosphoadenosine 5'-phosphosulfate binding
N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000462406.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000462406.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000462406.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000462406.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000476765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000476765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000476765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay PSR1000182880.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000182880.hg.1 ENST00000462406.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000182880.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182880.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182880.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000182880.hg.1 ENST00000462406.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000182880.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182880.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182880.hg.1 TC1000012117.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000346248.7 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000435907.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000462406.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000462406.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000462406.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000462406.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000476765.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000476765.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000476765.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 ENST00000628426.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270764.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_001270765.1 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_014863.3 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq
Affymetrix Human Clariom D Assay TC1000012117.hg.1 NM_015892.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0