This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01037828_m1
See other LIPN GE Assays ›
Gene Symbol:
LIPN
Gene Name:
lipase family member N
Gene Aliases:
ARCI8, LI4, LIPL4, bA186O14.3
Chromosome Location:
Chr.10: 88757226 - 88783662 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01037828_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIPN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001102469.1 NP_001095939.1 1 - 2 99 119
XM_005270049.3 XP_005270106.1 3 - 4 391 119
XM_011540083.2 XP_011538385.1 3 - 4 428 119
XM_011540084.2 XP_011538386.1 2 - 3 313 119
XM_011540085.2 XP_011538387.1 3 - 4 428 119
XM_011540086.2 XP_011538388.1 3 - 4 428 119
XM_017016549.1 XP_016872038.1 3 - 4 428 119
GenBank mRNA EF426483.1 - 1 - 2 99 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000008382.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000629.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

lipid catabolic process
lipid digestion
lipoprotein lipase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10006907.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10006907.hg.1 ENST00000491766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10006907.hg.1 NM_001102469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10006907.hg.1 TC10000629.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10006909.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10006909.hg.1 NM_001102469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10006909.hg.1 TC10000629.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10006908.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10006908.hg.1 ENST00000491766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10006908.hg.1 NM_001102469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10006908.hg.1 TC10000629.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156770.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000156770.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156770.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000156770.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156770.hg.1 TC1000008382.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000156771.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000156771.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156771.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000156771.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156771.hg.1 TC1000008382.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000156772.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000156772.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156772.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000156772.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156772.hg.1 TC1000008382.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008382.hg.1 ENST00000404459.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay TC1000008382.hg.1 NM_001102469.1 Refseq
Affymetrix Human Clariom D Assay TC1000008382.hg.1 NM_001102469.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z47m5:80/100.66.77.18:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0