This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01038327_m1
See other PARK2 GE Assays ›
Gene Symbol:
PARK2
Gene Name:
parkin RBR E3 ubiquitin protein ligase
Gene Aliases:
AR-JP, LPRS2, PDJ, PRKN
Chromosome Location:
Chr.6: 161347417 - 162727802 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01038327_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PARK2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004562.2 NP_004553.2 4 - 5 671 71
XM_011535863.1 XP_011534165.1 4 - 5 668 71
XM_017010908.1 XP_016866397.1 4 - 5 1585 71
XM_017010909.1 XP_016866398.1 3 - 4 465 71
GenBank mRNA AB009973.1 - 4 - 5 638 71
AB245403.1 - 4 - 5 629 71
AF381282.1 - 2 - 3 296 71
AF381283.1 - 2 - 3 373 71
AK292590.1 - 4 - 5 632 71
BC022014.2 - 4 - 5 635 4824892 71
GU345837.1 - 3 - 4 390 71
GU345840.1 - 3 - 4 554 71
GU361468.1 - 2 - 3 313 71
GU361469.1 - 4 - 5 651 71
GU361470.1 - 4 - 5 652 71
KC357595.1 - 6 - 7 749 71
KJ534911.1 - 3 - 4 537 71
KJ534912.1 - 3 - 4 537 71
KU178240.1 - 3 - 4 537 71
KU178241.1 - 3 - 4 537 71
KU178242.1 - 3 - 4 534 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600013800.hg
This assay targets the same location as HTA transcript cluster(s) TC06002286.hg

Additional Information:

This assay is in the 5'UTR region of the AF381282.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PARK2

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
protein polyubiquitination
mitochondrial fission
mitophagy
negative regulation of protein phosphorylation
startle response
transcription, DNA-templated
protein monoubiquitination
response to oxidative stress
mitochondrion organization
central nervous system development
learning
adult locomotory behavior
proteasomal protein catabolic process
regulation of autophagy
positive regulation of gene expression
negative regulation of gene expression
positive regulation of mitochondrial fusion
negative regulation of mitochondrial fusion
regulation of mitochondrion organization
regulation of glucose metabolic process
free ubiquitin chain polymerization
regulation of dopamine secretion
macroautophagy
protein ubiquitination
regulation of protein ubiquitination
protein destabilization
negative regulation of actin filament bundle assembly
regulation of lipid transport
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of glucokinase activity
cellular response to unfolded protein
response to endoplasmic reticulum stress
synaptic transmission, glutamatergic
protein K29-linked ubiquitination
ERAD pathway
regulation of dopamine metabolic process
norepinephrine metabolic process
dopamine metabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of DNA binding
negative regulation of neuron apoptotic process
cellular protein catabolic process
cellular protein metabolic process
protein K27-linked ubiquitination
negative regulation by host of viral genome replication
positive regulation of protein catabolic process
positive regulation of transcription from RNA polymerase II promoter
negative regulation of JNK cascade
negative regulation of insulin secretion
protein stabilization
positive regulation of neurotransmitter uptake
dopamine uptake involved in synaptic transmission
protein autoubiquitination
regulation of mitochondrial membrane potential
zinc ion homeostasis
negative regulation of cell death
regulation of canonical Wnt signaling pathway
parkin-mediated mitophagy in response to mitochondrial depolarization
neuron cellular homeostasis
protein K63-linked ubiquitination
protein localization to mitochondrion
aggresome assembly
protein K48-linked ubiquitination
protein K11-linked ubiquitination
cellular response to manganese ion
protein K6-linked ubiquitination
negative regulation of canonical Wnt signaling pathway
positive regulation of mitochondrial fission
negative regulation of release of cytochrome c from mitochondria
cellular response to toxic substance
mitophagy in response to mitochondrial depolarization
regulation of cellular response to oxidative stress
negative regulation of neuron death
positive regulation of proteasomal protein catabolic process
negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator
negative regulation of primary amine oxidase activity
positive regulation of protein linear polyubiquitination
regulation of synaptic vesicle transport
negative regulation of oxidative stress-induced cell death
regulation of protein targeting to mitochondrion
positive regulation of tumor necrosis factor-mediated signaling pathway
cellular response to dopamine
negative regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
positive regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
negative regulation of endoplasmic reticulum stress-induced neuron intrinsic apoptotic signaling pathway
positive regulation of dendrite extension
negative regulation of spontaneous neurotransmitter secretion
regulation of reactive oxygen species metabolic process
negative regulation of reactive oxygen species metabolic process
transcription regulatory region sequence-specific DNA binding
G-protein coupled receptor binding
transcription factor activity, sequence-specific DNA binding
actin binding
ubiquitin-protein transferase activity
protein binding
beta-catenin binding
zinc ion binding
tubulin binding
ligase activity
SH3 domain binding
enzyme binding
kinase binding
protein kinase binding
PDZ domain binding
Hsp70 protein binding
heat shock protein binding
ubiquitin conjugating enzyme binding
ubiquitin protein ligase binding
identical protein binding
histone deacetylase binding
ubiquitin binding
phospholipase binding
chaperone binding
ubiquitin protein ligase activity
cullin family protein binding
ubiquitin protein ligase activity involved in ERAD pathway
ubiquitin-specific protease binding
F-box domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000338468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000338468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000338468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000338468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000338468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000338468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000338468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000338468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000338468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000338468.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366892.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366892.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366892.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366892.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366892.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366892.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366892.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366892.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366892.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366892.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000366898.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000479615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000479615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000479615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000479615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000479615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000479615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000479615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000610470.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000610470.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000610470.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000610470.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000610470.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000612485.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000612485.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000612540.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000612540.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000612540.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000612540.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 ENST00000615065.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_004562.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013987.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013987.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013987.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013987.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013987.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013987.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013987.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013987.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013987.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013987.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013987.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013987.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013987.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013987.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013988.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013988.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013988.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013988.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013988.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013988.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013988.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013988.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013988.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013988.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013988.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013800.hg.1 NM_013988.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0