This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01039514_g1
See other NDOR1 GE Assays ›
Gene Symbol:
NDOR1
Gene Name:
NADPH dependent diflavin oxidoreductase 1
Gene Aliases:
NR1, bA350O14.9
Chromosome Location:
Chr.9: 137205667 - 137219361 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01039514_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDOR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144026.2 NP_001137498.1 1 - 2 246 77
NM_001144027.2 NP_001137499.1 1 - 2 246 77
NM_001144028.2 NP_001137500.1 1 - 2 246 77
NM_014434.3 NP_055249.1 1 - 2 246 77
XM_011518545.1 XP_011516847.1 1 - 2 236 77
XM_011518548.2 XP_011516850.1 1 - 2 225 77
XM_017014626.1 XP_016870115.1 1 - 2 246 77
XM_017014627.1 XP_016870116.1 1 - 2 246 77
GenBank mRNA AF199509.1 - 1 - 2 182 77
AK026089.1 - 1 - 2 218 77
AK074403.1 - 1 - 2 207 77
AK225995.1 - 1 - 2 217 77
AK290026.1 - 1 - 2 243 77
AY077845.1 - 1 - 2 164 77
BC015735.1 - 1 - 2 182 4869561 77
BC078668.1 - 1 - 2 182 77
BC093782.1 - 1 - 2 163 7939627 77
BC111943.1 - 1 - 2 163 8327411 77
DA408370.1 - 1 - 2 246 77
DA858250.1 - 1 - 2 246 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011518548.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDOR1

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

cell death
iron-sulfur cluster assembly
cellular response to menadione
oxidation-reduction process
NADPH-hemoprotein reductase activity
protein binding
FMN binding
oxidoreductase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-l2zpb:80/100.66.75.138:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0