This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01039823_m1
See other SLC25A16 GE Assays ›
Gene Symbol:
SLC25A16
Gene Name:
solute carrier family 25 member 16
Gene Aliases:
D10S105E, GDA, GDC, HGT.1, ML7, hML7
Chromosome Location:
Chr.10: 68479961 - 68527523 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01039823_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC25A16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001324312.1 NP_001311241.1 4 - 5 569 72
NM_001324313.1 NP_001311242.1 4 - 5 569 72
NM_001324314.1 NP_001311243.1 5 - 6 751 72
NM_001324315.1 NP_001311244.1 4 - 5 1426 72
NM_001324317.1 NP_001311246.1 3 - 4 435 72
NM_152707.3 NP_689920.1 4 - 5 569 72
XM_011540211.2 XP_011538513.1 4 - 5 569 72
GenBank mRNA AK290255.1 - 4 - 5 498 72
AK295287.1 - 4 - 5 534 72
AK298510.1 - 5 - 6 644 72
BC030266.1 - 4 - 5 520 5241247 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000010849.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001349.hg.1

Additional Information:

This assay is in the 5'UTR region of the NM_001324317.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC25A16

Panther Classification:

Molecular Function -

mitochondrial carrier protein

Gene Ontology Categories:

Function(s) Process(es)

translation
transport
coenzyme biosynthetic process
transmembrane transport
structural constituent of ribosome
secondary active transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000265870.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000439904.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000439904.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000474927.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000474927.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000474927.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000474927.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000491102.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000491102.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000491102.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000491102.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000491102.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000491102.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000493963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000608053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000608053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000608053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 ENST00000609923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010849.hg.1 NM_152707.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0