This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01039940_m1
See other ATXN1 GE Assays ›
Gene Symbol:
ATXN1
Gene Name:
ataxin 1
Gene Aliases:
ATX1, D6S504E, SCA1
Chromosome Location:
Chr.6: 16299112 - 16761490 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01039940_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATXN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000332.3 NP_000323.2 2 - 3 243 78
GenBank mRNA X79204.1 - 2 - 3 207 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600010921.hg
This assay targets the same location as HTA transcript cluster(s) TC06001293.hg

Additional Information:

This assay is in the 5'UTR region of the NM_000332.3,X79204.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATXN1

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
RNA processing
negative regulation of transcription, DNA-templated
nuclear export
DNA binding
protein binding
protein C-terminus binding
poly(U) RNA binding
poly(G) binding
identical protein binding
protein self-association

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR06015508.hg.1 ENST00000244769.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06015508.hg.1 ENST00000344707.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06015508.hg.1 ENST00000436367.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06015508.hg.1 ENST00000450222.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06015508.hg.1 ENST00000467008.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06015508.hg.1 NM_000332.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06015508.hg.1 NM_001128164.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06015508.hg.1 TC06001293.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 ENST00000467008.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 ENST00000479680.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 ENST00000492857.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 ENST00000495178.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 ENST00000244769.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 ENST00000436367.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 ENST00000467008.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 ENST00000473388.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 ENST00000479680.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 ENST00000492857.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 ENST00000495178.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 NM_000332.3 Refseq
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 NM_001128164.1 Refseq
Affymetrix Human Clariom D Assay PSR0600174176.hg.1 TC0600010921.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline