This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01040477_mH
See other RPL3L GE Assays ›
Gene Symbol:
RPL3L
Gene Name:
ribosomal protein L3 like
Gene Aliases:
-
Chromosome Location:
Chr.16: 1944571 - 1955123 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01040477_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL3L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005061.2 NP_005052.1 4 - 5 547 74
XM_011522571.2 XP_011520873.1 4 - 5 1007 74
GenBank mRNA AK313162.1 - 4 - 5 559 74
BC050413.1 - 4 - 5 539 6204847 74
U65581.1 - 4 - 5 548 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600009076.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000767.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPL3L

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal large subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
RNA binding
structural constituent of ribosome

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16011210.hg.1 ENST00000268661.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16011210.hg.1 NM_005061.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16011210.hg.1 TC16000767.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000565426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000565426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000565426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000565426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000566484.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000566484.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 ENST00000566484.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Clariom D Assay PSR1600168839.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600168839.hg.1 NM_005061.2 Refseq
Affymetrix Human Clariom D Assay PSR1600168839.hg.1 TC1600009076.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600168839.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600168839.hg.1 NM_005061.2 Refseq
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000268661.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000565426.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000565426.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000565426.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000565426.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000566484.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000566484.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 ENST00000566484.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Clariom D Assay TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Clariom D Assay TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Clariom D Assay TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Clariom D Assay TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Clariom D Assay TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Clariom D Assay TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Clariom D Assay TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Clariom D Assay TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Clariom D Assay TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Clariom D Assay TC1600009076.hg.1 NM_005061.2 Refseq
Affymetrix Human Clariom D Assay TC1600009076.hg.1 NM_005061.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline