| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000566484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000566484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000566484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011208.hg.1 |
ENST00000268661.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011208.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011208.hg.1 |
TC16000767.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1600168837.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168837.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168837.hg.1 |
TC1600009076.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600168837.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168837.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000566484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000566484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000566484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |