This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01040628_g1
See other AWAT2 GE Assays ›
Gene Symbol:
AWAT2
Gene Name:
acyl-CoA wax alcohol acyltransferase 2
Gene Aliases:
ARAT, DC4, DGAT2L4, MFAT, WS
Chromosome Location:
Chr.X: 70040542 - 70049970 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01040628_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AWAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002254.1 NP_001002254.1 2 - 3 202 62
XM_011530876.2 XP_011529178.1 2 - 3 234 62
GenBank mRNA AY605053.1 - 2 - 3 202 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00009952.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X001108.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AWAT2

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

wax biosynthetic process
acylglycerol acyl-chain remodeling
retinol metabolic process
cellular lipid metabolic process
2-acylglycerol O-acyltransferase activity
long-chain-alcohol O-fatty-acyltransferase activity
retinol O-fatty-acyltransferase activity
wax ester synthase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR0X015123.hg.1 TC0X001108.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X015123.hg.1 ENST00000276101.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X015123.hg.1 NM_001002254.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X015120.hg.1 TC0X001108.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X015120.hg.1 ENST00000276101.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X015120.hg.1 NM_001002254.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000440401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000440401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000440401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000440401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000440401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000440401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000440401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000443056.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000443056.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000443056.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000443056.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 ENST00000443056.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X015122.hg.1 TC0X001108.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X015122.hg.1 ENST00000276101.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X015122.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00167976.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00167976.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00167976.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00167976.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00167976.hg.1 TC0X00009952.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000440401.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000440401.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000440401.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000440401.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000440401.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000440401.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000440401.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000443056.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000443056.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000443056.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000443056.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 ENST00000443056.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009952.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00167973.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00167973.hg.1 ENST00000443056.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00167973.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00167973.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00167973.hg.1 ENST00000443056.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00167973.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00167973.hg.1 TC0X00009952.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00167975.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00167975.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00167975.hg.1 ENST00000276101.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00167975.hg.1 NM_001002254.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00167975.hg.1 TC0X00009952.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0