This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01040777_gH
See other NAXE GE Assays ›
Gene Symbol:
NAXE
Gene Name:
NAD(P)HX epimerase
Gene Aliases:
AIBP, APOA1BP, YJEFN1
Chromosome Location:
Chr.1: 156591762 - 156599818 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01040777_gH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAXE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144772.2 NP_658985.2 1 - 2 215 181
XM_017000319.1 XP_016855808.1 1 - 2 219 181
GenBank mRNA AJ315849.1 - 1 - 2 203 181
AK294835.1 - 1 - 2 223 181
AK298222.1 - 1 - 2 235 181
BC056917.1 - 1 - 2 235 3873464 181
BC100931.1 - 1 - 2 205 40003997 181
BC100932.1 - 1 - 2 203 40003998 181
BC100933.1 - 1 - 2 203 40003999 181
BC100934.2 - 1 - 2 203 40004000 181
BG723681.1 - 1 - 2 215 181
BQ430064.1 - 1 - 2 248 181

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100010185.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001323.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK294835.1,BC100932.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NAXE

Panther Classification:

Molecular Function -

epimerase/racemase isomerase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nicotinamide metabolic process
biological_process
protein homotetramerization
nucleotide binding
protein binding
protein homodimerization activity
metal ion binding
NADHX epimerase activity
NADPHX epimerase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500009651.hg.1 ENST00000512891.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020698.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020698.hg.1 ENST00000368234.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020698.hg.1 ENST00000368235.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020698.hg.1 ENST00000446584.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020698.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020698.hg.1 NM_144772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01020698.hg.1 TC01001323.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01020697.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020697.hg.1 ENST00000368234.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020697.hg.1 ENST00000368235.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020697.hg.1 ENST00000446584.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020697.hg.1 NM_144772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01020697.hg.1 TC01001323.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01020700.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020700.hg.1 ENST00000368234.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020700.hg.1 ENST00000368235.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020700.hg.1 ENST00000446584.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020700.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020700.hg.1 NM_144772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01020700.hg.1 TC01001323.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01020702.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020702.hg.1 ENST00000368234.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020702.hg.1 ENST00000368235.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020702.hg.1 ENST00000446584.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020702.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01020702.hg.1 NM_144772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01020702.hg.1 TC01001323.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000488840.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000488840.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000488840.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000488840.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 ENST00000488840.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay PSR0100174591.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100174591.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174591.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174591.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100174591.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay PSR0100174591.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100174591.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174591.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174591.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100174591.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay PSR0100174591.hg.1 TC0100010185.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100174587.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100174587.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174587.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174587.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay PSR0100174587.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100174587.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174587.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174587.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay PSR0100174587.hg.1 TC0100010185.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100174589.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100174589.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174589.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174589.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100174589.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay PSR0100174589.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100174589.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174589.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174589.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100174589.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay PSR0100174589.hg.1 TC0100010185.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000488840.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000488840.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000488840.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000488840.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 ENST00000488840.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay TC0100010185.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay TC0500009651.hg.1 ENST00000512891.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100174588.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100174588.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174588.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174588.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay PSR0100174588.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100174588.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174588.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174588.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay PSR0100174588.hg.1 TC0100010185.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100174593.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100174593.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174593.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174593.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100174593.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay PSR0100174593.hg.1 ENST00000368233.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100174593.hg.1 ENST00000368234.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174593.hg.1 ENST00000368235.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100174593.hg.1 ENST00000467374.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100174593.hg.1 NM_144772.2 Refseq
Affymetrix Human Clariom D Assay PSR0100174593.hg.1 TC0100010185.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100174592.hg.1 TC0100010185.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 178575 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline