This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01040830_g1
See other GLT8D1 GE Assays ›
Gene Symbol:
GLT8D1
Gene Name:
glycosyltransferase 8 domain containing 1
Gene Aliases:
AD-017, MSTP139
Chromosome Location:
Chr.3: 52694484 - 52706083 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01040830_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLT8D1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018446.3 NP_060916.1 2 - 3 686 110
GenBank mRNA AF157318.1 - 2 - 3 131 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300011182.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF157318.1,NM_018446.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GLT8D1

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

polysaccharide biosynthetic process
transferase activity, transferring glycosyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000266014.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000394783.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000407584.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000464705.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000478968.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000479553.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000484163.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000485899.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000487642.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000489119.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000491606.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000497436.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000497953.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 NM_001010983.2 Refseq
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 NM_001278280.1 Refseq
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 NM_001278281.1 Refseq
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 NM_018446.3 Refseq
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 NM_152932.2 Refseq
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000266014.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000394783.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000407584.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000464705.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000478968.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000479553.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000484163.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000485899.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000487642.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000489119.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000491606.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000497436.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 ENST00000497953.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 NM_001010983.2 Refseq
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 NM_001278280.1 Refseq
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 NM_001278281.1 Refseq
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 NM_018446.3 Refseq
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 NM_152932.2 Refseq
Affymetrix Human Clariom D Assay PSR0300183574.hg.1 TC0300011182.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0