This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01041061_m1
See other SMPDL3A GE Assays ›
Gene Symbol:
SMPDL3A
Gene Name:
sphingomyelin phosphodiesterase acid like 3A
Gene Aliases:
ASM3A, ASML3a, yR36GH4.1
Chromosome Location:
Chr.6: 122787180 - 122809720 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01041061_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMPDL3A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006714.4 NP_006705.1 1 - 2 406 60
XM_005266798.2 XP_005266855.1 1 - 2 406 60
XM_005266799.2 XP_005266856.1 1 - 2 406 60
XM_017010203.1 XP_016865692.1 1 - 2 406 60
XM_017010204.1 XP_016865693.1 1 - 2 105 60
GenBank mRNA AK000184.1 - 1 - 2 179 60
BC018999.2 - 1 - 2 176 3138813 60
DA510600.1 - 1 - 2 406 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600009333.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000950.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_017010204.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SMPDL3A

Panther Classification:

Molecular Function -

hydrolase phosphodiesterase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

sphingomyelin catabolic process
biological_process
nucleoside triphosphate catabolic process
sphingomyelin phosphodiesterase activity
protein binding
phosphoric diester hydrolase activity
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR06011065.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06011065.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06011065.hg.1 ENST00000539041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06011065.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06011065.hg.1 TC06000950.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06011062.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06011062.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06011062.hg.1 TC06000950.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06011064.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06011064.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06011064.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06011064.hg.1 TC06000950.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06011061.hg.1 NM_006714.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06011061.hg.1 TC06000950.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay TC0600009333.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay PSR0600163922.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600163922.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600163922.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600163922.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay PSR0600163922.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay PSR0600163922.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600163922.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600163922.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600163922.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay PSR0600163922.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay PSR0600163922.hg.1 TC0600009333.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600163918.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600163918.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay PSR0600163918.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay PSR0600163918.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600163918.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay PSR0600163918.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay PSR0600163918.hg.1 TC0600009333.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600163921.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600163921.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600163921.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600163921.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay PSR0600163921.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay PSR0600163921.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600163921.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600163921.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600163921.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay PSR0600163921.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay PSR0600163921.hg.1 TC0600009333.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600163923.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600163923.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600163923.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay PSR0600163923.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600163923.hg.1 ENST00000487215.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600163923.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay PSR0600163923.hg.1 TC0600009333.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600163919.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600163919.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600163919.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay PSR0600163919.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay PSR0600163919.hg.1 ENST00000368440.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600163919.hg.1 ENST00000539041.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600163919.hg.1 NM_001286138.1 Refseq
Affymetrix Human Clariom D Assay PSR0600163919.hg.1 NM_006714.4 Refseq
Affymetrix Human Clariom D Assay PSR0600163919.hg.1 TC0600009333.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline