| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011066.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011066.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011066.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011066.hg.1 |
TC06000950.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009333.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163924.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163924.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163924.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163924.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163924.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163924.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163924.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163924.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163924.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163924.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163924.hg.1 |
TC0600009333.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600163925.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163925.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163925.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163925.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163925.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163925.hg.1 |
ENST00000368440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163925.hg.1 |
ENST00000487215.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163925.hg.1 |
ENST00000539041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163925.hg.1 |
NM_001286138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163925.hg.1 |
NM_006714.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163925.hg.1 |
TC0600009333.hg.1 |
Clariom D TC |